missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Vismodegib Hedgehog inhibitor
May be dissolved in DMSO (200 mg/mL); or ethanol (10 mg/mL, warm)
138.00€ - 216.00€
Specifications
CAS | 879085-55-9 |
---|---|
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Donor Source | Synthetic |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18224844
|
Novus Biologicals™
NBP2-62758-5MG |
5 mg |
138.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18277543
|
Novus Biologicals™
NBP2-62758-25MG |
25 mg |
216.00€
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Purity:>98% (HPLC); NMR (conforms)
- Molecular Weight: 421.3
- PubChem ID:24776445
Specifications
879085-55-9 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Synthetic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Vismodegib Hedgehog inhibitor