missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Vismodegib Hedgehog inhibitor
May be dissolved in DMSO (200 mg/mL); or ethanol (10 mg/mL, warm)
Brand: Novus Biologicals™ NBP2-62758-25MG
This item is not returnable.
View return policy
Description
- Purity:>98% (HPLC); NMR (conforms)
- Molecular Weight: 421.3
- PubChem ID:24776445

Specifications
Off-white | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
25 mg |
879085-55-9 | |
Synthetic | |
Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Vismodegib Hedgehog inhibitor > 25 mg
Spot an opportunity for improvement?Share a Content Correction