missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Vismodegib Hedgehog inhibitor

May be dissolved in DMSO (200 mg/mL); or ethanol (10 mg/mL, warm)

Brand:  Novus Biologicals™ NBP2-62758-5MG

 View more versions of this product

Product Code. 18224844

  • 138.00€ / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Purity:>98% (HPLC); NMR (conforms)
  • Molecular Weight: 421.3
  • PubChem ID:24776445
TRUSTED_SUSTAINABILITY
Specifications

Specifications

879085-55-9
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Synthetic
5 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Vismodegib Hedgehog inhibitor > 5 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.