Learn More
SI Analytics™ ViscoSystem™ AVS 470 Viscometer
Modular viscometry system ideal for simple and complex systems from Schott
Brand:  SI Analytics™ 285415712
Product Code. 11944749
  • 18885.00€ / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Precise, flexible capillary viscometry system
  • Allows suction and pressure measurements completely independently of a PC
  • Maximum independence and flexibility
  • Allows set up of a measuring station that meets your requirements, even under difficult conditions, eg to monitor production or control quality in the polymer and mineral oil industry
  • All types of viscosity calculation are already integrated into the device
  • PS2 keyboard to enter additional data
  • A serial printer can be used to conveniently document your results
  • Optional optical or TC version ViscoPump module can be used to adapt the unit at any time
  • Use your existing accessories such as thermostats, stands or flow-through coolers with the AVS 470
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Viscometer
Automatic viscometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ ViscoSystem™ AVS 470 Viscometer > Automatic viscometer