missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Slip-Resistant General Purpose Tapes and Treads, Series 613

This mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

299.00€ - 1750.00€

Specifications

Adhesive Type Pressure Sensitive Adhesive
Color Black and Yellow Striped
For Use With (Application) Warning of Slip Prone Areas
Length (English) 60 ft.
View More Specs

Products 6
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
17158750
View Documents
3M™ Safety-Walk™
7100133325
18 x 0.051 m
373.00€
2 rolls
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
17198750
View Documents
3M™ Safety-Walk™
7100134978
18 x 0.152 m
446.00€
1 roll
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
17148750
View Documents
3M™ Safety-Walk™
7100133324
18.3 x 0.025 m
373.00€
4 rolls
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
17118760
View Documents
3M™ Safety-Walk™
7100143181
18.3 x 0.102 m
299.00€
1 roll
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
17128760
View Documents
3M™ Safety-Walk™
7100143241
18.3 x 0.305 m
904.00€
1 roll
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
17168750
View Documents
3M™ Safety-Walk™
7100133326
18.3 x 0.610 m
1750.00€
1 roll
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
 
Description

Description

  • Diagonal black/yellow striped tape provides early warning of slip prone areas
  • Extremely visible, anti-slip tape alerts guests or workers to a potential hazard
  • High quality abrasive particles to ensure long-lasting friction and visibility
  • High performance pressure-sensitive adhesive for easy application
  • NFSI High Traction Certification
Specifications

Specifications

Pressure Sensitive Adhesive
Black and Yellow Striped
Warning of Slip Prone Areas
60 ft.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Safety-Walk™ Slip-Resistant General Purpose Tapes and Treads, Series 613

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.