Learn More
3M™ Safety-Walk™ Slip-Resistant General Purpose Tapes and Treads, Series 613
This mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
Brand:  3M™ Safety-Walk™ 7100133325
 View more versions of this product
Product Code. 17158750
  • 373.00€ / 2 rolls
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Diagonal black/yellow striped tape provides early warning of slip prone areas
  • Extremely visible, anti-slip tape alerts guests or workers to a potential hazard
  • High quality abrasive particles to ensure long-lasting friction and visibility
  • High performance pressure-sensitive adhesive for easy application
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
18 x 0.051 m
60 ft.
2 Pk.
0.051 m
Black and Yellow Striped
Warning of Slip Prone Areas
18 m
2 in.
Pressure Sensitive Adhesive
Floor Tape