missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Slip-Resistant General Purpose Tapes and Treads, Series 613

Product Code. 17148750
Click to view available options
Dimensions (L x W):
18 x 0.051 m
18 x 0.152 m
18.3 x 0.025 m
18.3 x 0.102 m
18.3 x 0.305 m
18.3 x 0.610 m
Unit Size:
1 roll
2 rolls
4 rolls
This item is not returnable. View return policy

Product Code. 17148750

Brand: 3M™ Safety-Walk™ 7100133324

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

  • Diagonal black/yellow striped tape provides early warning of slip prone areas
  • Extremely visible, anti-slip tape alerts guests or workers to a potential hazard
  • High quality abrasive particles to ensure long-lasting friction and visibility
  • High performance pressure-sensitive adhesive for easy application
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 18.3 x 0.025 m
For Use With (Application) Warning of Slip Prone Areas
Length (English) 60 ft.
Length (Metric) 18.3 m
Packaging Quantity 4 Pk.
Width (English) 1 in.
Width (Metric) 0.025 m
Adhesive Type Pressure Sensitive Adhesive
Color Black and Yellow Striped
Type Floor Tape
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.