Learn More
Novus Biologicals™ Recombinant Human FGF-23 His Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
529.00€ - 779.00€
Specifications
Concentration | 0.25 mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol |
Gene ID (Entrez) | 8074 |
Molecular Weight (g/mol) | 26.4 kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18619060
|
Novus Biologicals™
NBP2-77508-100ug |
100 μg |
779.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18215197
|
Novus Biologicals™
NBP2-77508-0.05MG |
0.05 mg |
529.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A recombinant protein with a C terminal His-tag and corresponding to the amino acids 25-251 of Human FGF-23.
Source: Baculovirus
Amino Acid Sequence:
ADPYPNASPL LGSSWGGLIH LYTATARNSY HLQIHKNGHV DGAPHQTIYS ALMIRSEDAG FVVITGVMSR RYLCMDFRGN IFGSHYFDPE NCRFQHQTLE NGYDVYHSPQ YHFLVSLGRA KRAFLPGMNP PPYSQFLSRR NEIPLIHFNT PIPRRHTRSA EDDSERDPLN VLKPRARMTP APASCSQELP SAEDNSPMAS DPLGVVRGGR VNTHAGGTGP EGCRPFAKFI HHHHHH
Specifications
0.25 mg/mL | |
Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol | |
26.4 kDa | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
FGF23 | |
< 1.0 EU per 1 μg of protein (determined by LAL method) |
SDS-PAGE | |
8074 | |
Baculovirus | |
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol | |
>90% SDS-PAGE |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.