missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human FGF-23 His Protein

Product Code. 18215197 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
100 μg
Unit Size:
0.05mg
100µg
This item is not returnable. View return policy

Product Code. 18215197

Brand: Novus Biologicals™ NBP2775080.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C terminal His-tag and corresponding to the amino acids 25-251 of Human FGF-23.

Source: Baculovirus

Amino Acid Sequence:

ADPYPNASPL LGSSWGGLIH LYTATARNSY HLQIHKNGHV DGAPHQTIYS ALMIRSEDAG FVVITGVMSR RYLCMDFRGN IFGSHYFDPE NCRFQHQTLE NGYDVYHSPQ YHFLVSLGRA KRAFLPGMNP PPYSQFLSRR NEIPLIHFNT PIPRRHTRSA EDDSERDPLN VLKPRARMTP APASCSQELP SAEDNSPMAS DPLGVVRGGR VNTHAGGTGP EGCRPFAKFI HHHHHH

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol
Gene ID (Entrez) 8074
Molecular Weight (g/mol) 26.4 kDa
Quantity 0.05 mg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Alias ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor
Gene Symbol FGF23
Buffer Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity or Quality Grade >90% SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.