missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human FGF-23 His Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Brand:  Novus Biologicals™ NBP2-77508-100ug

 View more versions of this product

Product Code. 18619060

  • 779.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A recombinant protein with a C terminal His-tag and corresponding to the amino acids 25-251 of Human FGF-23.

Source: Baculovirus

Amino Acid Sequence:

ADPYPNASPL LGSSWGGLIH LYTATARNSY HLQIHKNGHV DGAPHQTIYS ALMIRSEDAG FVVITGVMSR RYLCMDFRGN IFGSHYFDPE NCRFQHQTLE NGYDVYHSPQ YHFLVSLGRA KRAFLPGMNP PPYSQFLSRR NEIPLIHFNT PIPRRHTRSA EDDSERDPLN VLKPRARMTP APASCSQELP SAEDNSPMAS DPLGVVRGGR VNTHAGGTGP EGCRPFAKFI HHHHHH

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.25 mg/mL
Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol
26.4 kDa
Baculovirus
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor
Phosphate Buffered Saline (pH 7.4), 2 mM DTT, 1 mM EDTA, 10% glycerol
>90% SDS-PAGE
SDS-PAGE
8074
100 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
FGF23
< 1.0 EU per 1 μg of protein (determined by LAL method)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Recombinant Human FGF-23 His Protein > 100 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.