missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTM Technologies™ Multi-Operator Fume Hoods
Used for filtration where extraction arms or other accessories are needed. FTM Technologies™ Multi-Operator Fume Hoods provide multi-workstation filtration for welding fumes, solvents and other hazardous substances.
1530.00€ - 1615.00€
Specifications
Dimensions (L x W x H) | 350 x 200 x 430 mm |
---|---|
Flow Rate | 50 to 450 m3/h |
Hertz | 50 Hz |
Voltage | 230 V |
Weight (Metric) | 6 kg |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11778044
|
FTM Technologies™
HI4P4S |
4 extraction inputs |
1530.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11778054
|
FTM Technologies™
HI4P6S |
6 extraction inputs |
1615.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Available with either 4 or 6 extraction inputs
- Delivered with ringed PVC pipe
- Compatible with the use of prefilters
- Can be used as a recirculating (ductless) fume hood, using active carbon filtration or dust filter, or with an external output (external filter plus exhaust duct)
Specifications
350 x 200 x 430 mm | |
50 Hz | |
6 kg |
50 to 450 m3/h | |
230 V |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FTM Technologies™ Multi-Operator Fume Hoods