Learn More
FTM Technologies™ Multi-Operator Fume Hoods
Used for filtration where extraction arms or other accessories are needed. FTM Technologies™ Multi-Operator Fume Hoods provide multi-workstation filtration for welding fumes, solvents and other hazardous substances.
Brand:  FTM Technologies™ HI4P6S
 View more versions of this product
Product Code. 11778054
  • 1615.00€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • Available with either 4 or 6 extraction inputs
  • Delivered with ringed PVC pipe
  • Compatible with the use of prefilters
  • Can be used as a recirculating (ductless) fume hood, using active carbon filtration or dust filter, or with an external output (external filter plus exhaust duct)
Specifications
Specifications
350 x 200 x 430 mm
50 Hz
230 V
50 to 450 m3/h
6 extraction inputs
6 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
FTM Technologies™ Multi-Operator Fume Hoods > Includes: 6 extraction inputs