Learn More
FTM Technologies™ Multi-Operator Fume Hoods
Used for filtration where extraction arms or other accessories are needed. FTM Technologies™ Multi-Operator Fume Hoods provide multi-workstation filtration for welding fumes, solvents and other hazardous substances.
Brand: FTM Technologies™ HI4P4S
This item is not returnable.
View return policy
Description
- Available with either 4 or 6 extraction inputs
- Delivered with ringed PVC pipe
- Compatible with the use of prefilters
- Can be used as a recirculating (ductless) fume hood, using active carbon filtration or dust filter, or with an external output (external filter plus exhaust duct)
Specifications
350 x 200 x 430 mm | |
50 Hz | |
230 V |
50 to 450 m3/h | |
4 extraction inputs | |
6 kg |
Product Suggestions
Customers who viewed this item also viewed
Multi-operator hood - HI4P4S
File size: 329.3 kB
Hotte multiposte - HI4P4S
File size: 326.6 kB
Product Content Correction
Product Title
FTM Technologies™ Multi-Operator Fume Hoods > Includes: 4 extraction inputs