Learn More
WTW™ Iron Test Kits
Brand: WTW™ 250435
255.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description

Specifications
For waste water, drinking water, foodstuffs | |
Certificates | |
180 Tests | |
Zinc (Zn) cell test kit |
For PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex, Photoflex photometers | |
180 | |
Photometric |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- iron test method
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : ammonium thioglycolate, thioglycolic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.