Learn More
Lovibond™ IRON (II) LR tablets
The suitable tablet for the fast result
Brand:  Lovibond™ 515420BT
Product Code. 15311845
  • 93.20€ / Pack of 100
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The suitable tablet for the fast result
Water analysis couldn't be easier. Precise dosing, long shelf life and easy handling make the tablet reagent the most popular indicator system and the cheapest way to have analyse a parameter.
The tablet reagent in the aluminum blister pack withstands almost all climatic conditions and external influences. In addition, there is no safety risk for the user.
Specifications
Specifications
100 Tablets
IRON(II) 0.02 to 1.0 mg/L
Reagent
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Iron II LR
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
SDS
Documents
Documents
Brochures
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lovibond™ IRON (II) LR tablets >