Learn More
Speirs Robertson™ Instrument Isolation Platform
Product Code. 11969434
Encompass_Preferred
Click to view available options
:
Dimensions (L x W): 30 x 45cm
Product Code. 11969434
Brand: Speirs Robertson™ AMDSR 90x75SS
This item requires a price update prior to being ordered.
Request a Price for this item here.
Improve performance of sensitive instruments by shielding them from vibrations. Speirs Robertson™ Instrument Isolation Platform is simple to install, easy to use, and maintenance free.
  • Sorbothane isolators
  • Improve instrument performance by removing vibrations
  • Suitable for instruments weighing up to 25kg
  • Combination of isolators and sound-deadening, layered construction
  • Seamless pharma-grade (Ra finish 0.3) stainless steel worktop
  • Chemically resistant and easy to clean