Learn More
Novus Biologicals™ INSTA-Blot Human Tissues
Applications: Western Blot
Brand: Novus Biologicals™ NBP2-30113
Description
- Screening proteins from various human tissues
- Contains 11 human tissue lysates (20μg total protein per lane) on 4-20% Tris-Glycine mini gel
- Prepared from resolution by SDS-PAGE at 125V for 1 hour
- Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
- Stained with amido black for visualization of proteins
Recommended Storage
Store at room temperature

Specifications
Tissues are from donors with no known disease. Tissues are lysed in lysis buffer (10 mM Tris, pH 8.0, 130 mM NaCl, 1% Triton X-100, 10 mM NaF, 10 mM NaPi, 10 mM NaPPi) containing Protease Inhibitor Cocktail (PIC) and PMSF. Prior to SDS-PAGE the tissue lysate is resuspended 1:1 with 2X SDS sample buffer (120 mM Tris-Hcl [pH 6.8], 20 mM EDTA, 4% SDS, 0.06% Bromophenol Blue, 20% glycerol, 0.4% B-mercaptoethanol). Use in western blot reported in scientific literature (PMID: 24704276) . | |
Western Blot | |
1 Membrane | |
PVDF |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.