Learn More
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials and Surfaces
Click to view available options
Dimensions (LxW):
101.6 x 101.6 mm
22.2 x 22.2 mm
23.9mm x 12.7mm + 11.1 mm
25.4 x 12.7 mm
25.4 x 25.4 mm
31.8 x 22.2 mm
50.8 x 25.4 mm
50.8 x 6.4 mm
Diameter (Metric):
25.4 mm
76.2 mm
Labels per Row:
1 Label
2 Labels
Unit Size:
Pack of 1000
Pack of 2000
Pack of 300
Pack of 3000
Pack of 4000
Pack of 500
Pack of 5000
Description
- Ultra-resistant adhesive ensures no flagging or detachment when stored in liquid nitrogen
- Thermal-transfer printable for the best printout and barcode quality
- Ideal for labeling ambient temperature as well as frozen surfaces without thawing first
- No overlap or wrap-around required for proper adhesion
- Wide temperature range resistance
Specifications
Specifications
| Finish | Medium Gloss |
| Format | Roll |
| Material | Thermoplastic Film |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| Sterility | Non-sterile |
| Labels per Row | 1 Label |
| Quantity | 1000/Pk. |
| Shape | Rectangle |
| Temperature Range (English) | -321°F to +250°F |
| Show More |