Learn More
Human CEACAM-16 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems FAB9885R-100UG
Description
CEACAM16 Monoclonal antibody specifically detects CEACAM16 in Human samples. It is validated for Flow Cytometry
Specifications
CEACAM16 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
carcinoembryonic antigen-related cell adhesion molecule 16, CEAL2, DFNA4B | |
Chinese Hamster Ovary cell line CHO-derived human CEACAM-16, Met1-Gly425, Accession # Q2WEN9 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2747B | |
Flow Cytometry | |
Q2WEN9 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
388551 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.