Learn More
SI Analytics™ pH Combination Electrode
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ pH Combination Electrode is robust, durable, and easy to clean.
Brand:  SI Analytics™ 285100231
 View more versions of this product
Product Code. 10040171
  • 368.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Proven high-end electrodes for demanding measurement tasks
  • Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
  • Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
  • Each pH combination electrode comes with individual serial number and quality certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plug Head
H-Glass
pH Combination Electrode
Sphere
0°C to 100°C
12 mm
Ground Joint Diaphragm
0 to 14
Silamid™
H 64
170 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ pH Combination Electrode > LengthMetric: 170mm