Learn More
BINDER™ Reinforced Rack
Reinforced shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
Brand: BINDER™ 8012-2414
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Specifications
KB 720, KB PRO 720, KBF 720, KBF PRO 720 | |
Stainless Steel | |
Rack |