Learn More
BINDER™ Reinforced Rack
Reinforced shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
291.00€ - 682.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
30238242
|
BINDER™
8012-2412 |
KBF 130, KBF PRO 130 |
291.00€
Each |
In Stock
Log in to see stock. |
||||||
30238289
|
BINDER™
8012-2413 |
KB 260, KB 470, KB PRO 260, KB PRO 470, KBF 260, KBF 470, KBF PRO 260, KBF PRO 470 |
402.00€
Each |
In Stock
Log in to see stock. |
||||||
30238381
|
BINDER™
8012-2414 |
KB 720, KB PRO 720, KBF 720, KBF PRO 720 |
581.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
30238196
|
BINDER™
8012-2415 |
KB 1060, KB PRO 1060, KBF 1060, KBF PRO 1060 |
682.00€
Each |
In Stock
Log in to see stock. |
||||||
Description
- Shelf rack made of stainless steel
- Includes set of U-rails
Specifications
Stainless Steel | |
Rack |
Product Title
BINDER™ Reinforced Rack