Learn More
Macherey-Nagel™ Duotest™ pH Paper Refill
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Refills give the user the best possible accuracy in pH measurement.
Brand: Macherey-Nagel™ 90314
This item is not returnable.
View return policy
Description
- Provides rapid and reliable results
- Available for a range of pH gradations
- Shows two different colors for each pH-value at intervals of 0.3 to 1 pH unit
- Allows more accurate reading and good estimation of intermediate values
Specifications
5 to 8 | |
Refill | |
3 Pack | |
pH Paper |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Macherey-Nagel™ Duotest™ pH Paper Refill > pH Range: 5 to 8