missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Duotest™ pH Paper Refill

Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Refills give the user the best possible accuracy in pH measurement.

34.45€

Specifications

Format Refill
Quantity 3 Pack
Type pH Paper
View More Specs

Products 4
Product Code Brand pH Range Price Quantity & Availability  
Product Code Brand pH Range Price Quantity & Availability  
10004700
View Documents
Macherey-Nagel™
90313
3.5 to 6.8
34.45€
Pack of 3
Please to purchase this item. Need a web account? Register with us today!
10499860
View Documents
Macherey-Nagel™
90315
7 to 10
34.45€
Pack of 3
Please to purchase this item. Need a web account? Register with us today!
10799671
View Documents
Macherey-Nagel™
90311
1 to 12
34.45€
Pack of 3
In Stock
to see stock.
Add to basket
 
10666561
View Documents
Macherey-Nagel™
90314
5 to 8
34.45€
Pack of 3
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Provides rapid and reliable results
  • Available for a range of pH gradations
  • Shows two different colors for each pH-value at intervals of 0.3 to 1 pH unit
  • Allows more accurate reading and good estimation of intermediate values
Specifications

Specifications

Refill
3 Pack
pH Paper
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Duotest™ pH Paper Refill

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.