Learn More
Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution
For routine hemostasis testing
Brand:  Greiner Bio-One 454332
 View more versions of this product
Product Code. 15402550
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • 13 × 75mm tubes filled with buffered tri-sodium citrate solution
  • 3.2 % (0.109 mol/L) concentration
  • Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • 18-month shelf life from date of manufacture
  • Blue nonscrew safety pull cap can be removed with a gentle pull
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sodium Citrate Blood Collection Tube
PET
Blue with Black Ring
3.2% Citrate Solution
3.5 mL
13 x 75 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution > 3.2% Citrate Solution; Fill volume: 3.5L