missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Coagulation Tubes with Sodium Citrate Solution
For routine hemostasis testing
44.09€ - 259.00€
Specifications
Product Type | Sodium Citrate Blood Collection Tube |
---|---|
Additive | 3.2% Citrate Solution |
Material | PET |
Tube Size | 13 x 75 mm |
Product Code | Brand | Additive | Closure Color | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Additive | Closure Color | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||
15402550
|
Greiner Bio-One
454332 |
3.2% Citrate Solution | Blue with Black Ring | 3.5 mL | Sodium Citrate Blood Collection Tube | N/A | |||
13078692
|
Greiner Bio-One
454322 |
3.2% Citrate Solution | Blue with White Ring | 1.8 mL | Sodium Citrate Blood Collection Tube |
259.00€
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- 13 × 75mm tubes filled with buffered tri-sodium citrate solution
- 3.2 % (0.109 mol/L) concentration
- Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- 18-month shelf life from date of manufacture
- Blue nonscrew safety pull cap can be removed with a gentle pull
Specifications
Sodium Citrate Blood Collection Tube | |
3.2% Citrate Solution | |
PET | |
13 x 75 mm |