missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
30.00€ - 104.00€
Specifications
Certifications/Compliance | DIN 38408 G4, EN ISO 7393 |
---|---|
Volume (Metric) | 5 mL |
Product Code | Brand | Range | Type | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Type | For Use With (Equipment) | Price | Quantity & Availability | |||
11773177
|
WTW™
250419 |
0.03 to 6.00 Cl2, free | Cell Test Kit | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
104.00€
Pack of 150 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
11783177
|
WTW™
250420 |
0.03 to 6.00 Cl2, free and total | Cell Test Kit | PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter |
104.00€
Pack of 150 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
11721866
|
WTW™
251414 |
0.02 to 2.00 mg/L Cl2, total | Powder Pillows Kit | PhotoLab™ 6000 photometer, Photoflex™ meter |
30.00€
Pack of 100 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
11701866
|
WTW™
251401 |
0.02 to 2.00 mg/L Cl2, free | Powder Pillows Kit | PhotoLab™ 6000 photometer, Photoflex™ meter |
30.50€
Pack of 100 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
10027842
|
WTW™
252012 |
0.010 to 6.00 Cl2, free and total | Reagent Test Kit | PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer |
87.50€
Each |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
11711866
|
WTW™
251402 |
0.5 to 5.0 mg/L Cl2, free | Powder Pillows Kit | PhotoLab™ 6000 photometer, Photoflex™ meter |
30.50€
Pack of 100 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
Description
Specifications
DIN 38408 G4, EN ISO 7393 | |
5 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- chlorine cell test (free chlorine)
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : boric acid, ethylenedinitrilotetraaceticacid, tetrasodium salt