Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 251414
 View more versions of this product
Product Code. 11721866
  • 30.00€ / Pack of 100
Estimated Shipment: 10-10-2024
to see stock.
Explore more special offers
Add to basket
Specifications
DIN 38408 G4, EN ISO 7393
Disinfectants, wastewater, drinking water
0.02 to 2.00 mg/L Cl2, total
4-Dimethylaminobenzaldehyde
5 mL
PhotoLab™ 6000 photometer, Photoflex™ meter
15°C to 25°C
Powder Pillows Kit