Learn More
WTW™ Chlorine Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 250420
 View more versions of this product
Product Code. 11783177
  • 110.00€ / Pack of 150
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
DIN 38408 G4, EN ISO 7393
Disinfectants, wastewater, drinking water
Free and total Chlorine
0.03 to 6.00 Cl2, free and total
Cell Test Kit
5 mL
PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer, Photoflex™ meter
150 Pack
DPD
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Chlorine Cell Test (free and total chlorine) Method: photometric, DPD 0.03 - 6.00 mg/l Cl
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : boric acid, Ethylenedinitrilotetraaceticacid,Tetrasodiumsalt