missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Chlorine Test Kits

Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits

Brand:  WTW™ 252012


 View more versions of this product

Product Code. 10027842

  • 87.50€ / Each
Estimated Shipment: 10-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN 38408 G4, EN ISO 7393
Disinfectants, wastewater, drinking water
Silicate
15°C to 25°C
Reagent Test Kit
5 mL
PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer
0.010 to 6.00 Cl2, free and total
Silicomolybdenum blue
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • chlorine test, free and total chlorine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : boric acid, ethylenedinitrilotetraaceticacid, tetrasodium salt

SDS
Documents

Documents

Certificates
Promotions

Promotions