Learn More
Tocris Bioscience™ CASIN
Cdc42 inhibitor
Brand:  Tocris Bioscience™ 5050/10
 View more versions of this product
Product Code. 18709957
  • 300.00€ / 10mg
Explore more special offers
Description
Description
CASIN is a Cdc42 GTPase inhibitor (IC50 = 2 μM). Reduces active levels of Cdc42 in aged hematopoietic stem cells (HSCs) to that in young HSCs. Reverses the aging-related and polarity phenotype of aged HSCs to that of young HSCs in vivo. Increases histone H4K16 acetylation in aged HSCs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
>98%
CASIN
Rho GTPase Inhibitors
10 mg
306.4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Tocris Bioscience™ CASIN > 10mg