missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CASIN
GREENER_CHOICE

Product Code. p-5545735
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18709957

Brand: Tocris Bioscience™ 5050/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cdc42 inhibitor

CASIN is a Cdc42 GTPase inhibitor (IC50 = 2 μM). Reduces active levels of Cdc42 in aged hematopoietic stem cells (HSCs) to that in young HSCs. Reverses the aging-related and polarity phenotype of aged HSCs to that of young HSCs in vivo. Increases histone H4K16 acetylation in aged HSCs.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type CASIN
Purity >98%
Molecular Weight (g/mol) 306.4
Inhibitors Rho GTPase Inhibitors

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.