missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ pHotoFlex™ Portable Photometer with pH and Turbidity
A unique combination of photometric, electro-chemical pH and turbidity measurement with lab precision
Brand: WTW™ 251110
3110.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
With its unique combination of photometric, electro-chemical pH and turbidity measurement with lab precision, the pHotoFlex™ Turb is the most versatile meter for extensive work - from the environmental and process monitoring at changing sites, via fish hatcheries all the way to drinkingn water analytics and monitoring by health offices and service labs. The energy-efficient LED optics with 6 wavelengths and a smart adapter for a variety of applications with 16 and 28 mm cuvettes allows the measurement of the smallest concentration with more than 150 programs for standard parameters, special methods and coloration. There is a large selection of test sets of lot-certified cuvettes all the way to economic powder reagents available. A list of favorites for the most commonly used tests, sample ID numbers and more than 3,000 measurements per battery set facilitate field measurements. The integrated pH function is a complete electro-chemical pH measurement of pH 0-16 with automatic buffer recognition (TEC/NIST) by using the comprehensive WTW selection of pH measuring chains. The temperature compensation automatically takes place in the admissible measuring ranges from -5 … 100 °C. The WTW MultiCal™ routine allows the automatic calibration with up to 3 calibration points. or field use, there is, for instance, the maintenance-free SenTix™ 41 is recommended, for precision measurements in the lab, the glass electrode SenTix™ 81 is frequently used. The pHotoFlex™Turb also has an infrared light source (IR) for the nephelometric turbidity measurement (90°) according to DIN 27027 / ISO 7027. In combination with th e long-term stable AMCO Clear™ standard, the highest possible precision is also warranted for the sensitive drinking water area. The optional PC software LSdata offers a GLP-compliant data evaluation with data filter and allows the input of up to 100 user-defined programs, automatically calculate these as methods and save them to the pHotoFlex™. For the convenient use in the field, there is a case set available, including the pH probe SenTix™ 41 and turbidity standards as a mobile lab with work surface and storage compartment as well as a lot of accessories, such as pH standards, tripod, pipettes and replacement batteries. For lab use, the photometer can be used in conjunction with a LabStation (optional) with site-supplied power and fitted with an external barcode reader.
Specifications
LED | |
4.62 in. | |
9.25 in. | |
234.9 mm | |
85.5 mm |
Environmental Monitoring, Water Treatment, Beverage Industry, Wine Industry, Process Control, Multiparameter Applications for Photometry, pH and Turbidity | |
117.3 mm | |
Full electro-chemical pH functions and integrated IR turbidity measurement acc. DIN/ISO with approx. 200 programs and 100 user-defined programs, 1000 data sets | |
436, 517, 557, 594, 610, 690, 860nm | |
3.37 in. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
WTW141 F2430 AG photoflex wtw ba75491f02 pHotoFlex
File size: 1.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ pHotoFlex™ Portable Photometer with pH and Turbidity > Dimensions: 234.9L x 117.3mmH
Spot an opportunity for improvement?Share a Content Correction