Learn More
WTW™ Calibration and Maintenance Set
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
85.10€ - 119.00€
Specifications
Type Calibration and Maintenance Set
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
10398683
View Documents
WTW™
108825
pH-3 Technical buffer solution 250mL (pH 4.01, pH 7, pH 10.01) trace, 2 bottles of 250mL of KCl 3mol/L bottles
85.10€
Each
In Stock
to see stock.
Edge
Add to basket
10634471
View Documents
WTW™
109415
With standard (DIN/NBS) buffer solution, 3 bottles, 250mL each of pH 4.006 - 6.865 - 9.180, 1 bottle with 250mL pepsin cleaning solution, 1 bottle with 250mL KCl solution 3M/L
119.00€
1 set
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Description
Description
  • Adapted to different electrode types
  • Cost-effective combinations
  • Calibration and maintenance in one assortment
Specifications
Specifications
Calibration and Maintenance Set
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
WTW™ Calibration and Maintenance Set