Learn More
WTW™ Calibration and Maintenance Set
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
Brand:  WTW™ 108825
83.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10398683
  • 85.10€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Adapted to different electrode types
  • Cost-effective combinations
  • Calibration and maintenance in one assortment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration and Maintenance Set
pH-3 Technical buffer solution 250mL (pH 4.01, pH 7, pH 10.01) trace, 2 bottles of 250mL of KCl 3mol/L bottles
Gel electrodes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Calibration and Maintenance Set > For use with: Gel Electrodes; Includes: pH-3 Technical buffer solution 250mL trace, 2 bottles of 250mL of KCl 3mol/L bottles