Learn More
WTW™ Calibration and Maintenance Set
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
Brand:  WTW™ 109415
116.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10634471
  • 119.00€ / 1 set
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Adapted to different electrode types
  • Cost-effective combinations
  • Calibration and maintenance in one assortment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration and Maintenance Set
With standard (DIN/NBS) buffer solution, 3 bottles, 250mL each of pH 4.006 - 6.865 - 9.180, 1 bottle with 250mL pepsin cleaning solution, 1 bottle with 250mL KCl solution 3M/L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
WTW™ Calibration and Maintenance Set > Includes: With standard (DIN/NBS) buffer solution, 3 bottles, 250mL each of pH 4.006 - 6.865 - 9.180, 1 bottle with 250mL pepsin cleaning solution, 1 bottle with 250mL KCl solution 3M/L