Learn More
purine-rich element binding protein A, Mouse, Clone: 1D6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PURA.
Brand: Abnova H00005813-M07.100ug
Description
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia. [provided by RefSeq
Sequence: TQGQTIALPAQGLIEFRDALAKLIDDYGVEEEPAELPEGTSLTVDNKRFFFDVGSNKYGVFMRVSEVKPTYRNSITVPYKVWAKFGHTFCKYSEETKKIQEKQREKRAACSpecifications
purine-rich element binding protein A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005859 | |
PURA | |
PURA (NP_005850, 183 a.a. ∼ 292 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Cell Cycle | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
1D6 | |
Mouse monoclonal antibody raised against a partial recombinant PURA. | |
PURA | |
PUR-ALPHA/PUR1/PURALPHA | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
5813 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.