Learn More
Thermo Scientific™ SOLA™ SPE Cartridges
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Brand:  Thermo Scientific™ 60109-002
196.30 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11869163
  • 234.58€ / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Description
Grow your laboratory productivity. Thermo Scientific™ SOLA™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
SOLA products are available in a 10mg/1mL cartridge format. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Confidence in results
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 mL
10 mg
SOLA
SPE Cartridge
SOLA SCX SPE Cartridge
Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Basic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Mixed Mode Strong Cation Exchanger, Polymeric
1 mL
Videos
  • Thermo Scientific - SOLA SPE cartridges and plates
    Thermo Scientific SOLA products revolutionise Solid Phase Extraction (SPE). This first fritless SPE product range provides greater reproducibility with cleaner, more consistent extracts.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific SOLA SPE cartridges and plates
    • Thermo Scientific - SOLA SPE cartridges and plates
      Thermo Scientific SOLA products revolutionise Solid Phase Extraction (SPE). This first fritless SPE product range provides greater reproducibility with cleaner, more consistent extracts.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific SOLA SPE cartridges and plates
      Promotions
      • Multiple Manufacturers
        Use promotion code 24138 to get amazing savings on products for your analysis
        Promo Code: 24138
        Expires2025-04-30
        Days
        22
        Hours
        22
        Minutes
        24
        Seconds
        55
      • Multiple Manufacturers
        Explore our new promotion and get up to 70% off some of our most popular products.
        Promo Code: 24127
        Expires2025-05-31
        Days
        53
        Hours
        22
        Minutes
        24
        Seconds
        55
      Product Content Correction
      Product Title
      Thermo Scientific™ SOLA™ SPE Cartridges > 10mg/1mL; SOLA SCX SPE cartridge; 100 pack
      Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.