Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Empty Well Plates
These empty 96-well plates are used with Thermo Scientific™ HyperSep 96-well plate applications.
Brand: Thermo Scientific™ 60300-319
23.80 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format.
- Available in a wide range of chemistries and bed weights
- Ideal for large samples and conform to industry standard configurations
- Allows for efficient routine processing by vacuum, by positive pressure or centrifugation.
- The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility.
- The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation.
- Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing.
- HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
2 mL | |
Solid Phase Extraction | |
96-well SPE Plate |
Product Suggestions
Customers who viewed this item also viewed
Promotions
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%