Learn More
KGW Isotherm™ Dewar Cylindrical Insulated Flasks
For the storage and insulation of temperature-sensitive goods such as dry ice, liquid nitrogen and other coolants. KGW Isotherm™ Dewar Cylindrical Insulated Flasks feature a robust design made from borosilicate glass 3.3.
Brand: KGW Isotherm™ 10214
This item is not returnable.
View return policy
Description
- Easy handling
- Cooling of small samples
- Fully silvered as standard
- Flexible applications in laboratory
- Available with or without side grip
Specifications
Blue | |
Cooling of small samples, storage of liquid nitrogen, dry ice, other coolants and as insulating flask | |
300mm | |
1500 mL | |
Borosilicate Glass |
115mm | |
245mm | |
No side grip | |
90 mm |
Product Content Correction
Product Title
KGW Isotherm™ Dewar Cylindrical Insulated Flasks > Capacity: 1500mL; Includes: No side grip; Inner Height: 245mm; Inner Diameter: 90mm