Learn More
Knick Elektronische™ 766 Waterproof Laboratory pH Meter
The 766 Waterproof Laboratory pH Meter is designed for standard applications in everyday lab routines. It combines practical functionality and easy operation with comprehensive safety functions.
Brand:  Knick Elektronische™ 766
Product Code. 10518181
  • 1700.00€ / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Gaincheck automatic device test
  • Trueline calibrated analog recorder output
  • Sensor monitoring with Sensoface icon alerts
  • Automatic calibration with patented Calimatic
  • EMC according to NAMUR
  • Simultaneous pH and temperature display
  • Simple operation
  • Leak-proof membrane keyboard
  • Robust housing
  • Protection category IP 54
  • 3-year warranty
gaincheck, Calimatic, record, recorders, pH, meter
Specifications
Specifications
Sensoface Icon Alerts, Automatic Calibration, Simultaneous pH and Temperature Display
Leak-proof membrane keyboard
Laboratory pH Meter
Standard applications in everyday lab routines
766 Laboratory pH Meter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Knick Elektronische™ 766 Waterproof Laboratory pH Meter > 766 Waterproof Laboratory pH Meter; Includes: Sensoface Icon Alerts, Automatic Calibration, Simultaneous pH and Temperature Display