Learn More
Thermo Scientific™ TSB Blood Bank Refrigerators
The TSB Series blood bank refrigerators are built with the security, quality, and environmentally sustainable features you have come to expect from Thermo Scientific.
Brand:  Thermo Scientific™ TSB1500REGBV
 View more versions of this product
Product Code. 17466202
  • 13266.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The Thermo Scientific TSB Series Blood Bank Refrigerators are designed to meet the strict requirements for storage of blood and blood related products in full compliance with 93/42/CE – class IIa regulation.
TSB Series Blood Bank Refrigerators are purpose built to meet the stringent guidelines to meet 93/42/CE class IIa regulation as a medical device for storage of blood and blood related products.
  • Wide range of sizes from 140L undercounter to 1500L double door.
  • Touchscreen digital controller with onboard datalogging
  • Whisper quite operation < 50 dBA
  • Triple pane glass doors.
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low energy consumption with real time energy monitoring
  • Low heat output
  • Rear access port
  • Electronic lock with password protection
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ TSB Blood Bank Refrigerators >