Learn More
Kautex™ Plastic, Jerry Can
Color: Natural
Brand: Kautex™ 2000090643
Description
- Plastic
- Containers
- Without caps
Specifications
2.6 gal. | |
Natural | |
Jerry Can |
10 L | |
Plastic |
Kautex - Expertise in packaging - Standard plastics
File size: 1.3 MB
Product Content Correction
Product Title
Kautex™ Plastic, Jerry Can > Capacity: 10L