Learn More
YSI™ Pro Series DO Cap Membrane Kit
71.70€
Specifications
Type | Membrane Kit |
---|---|
For Use With (Application) | For pro series galvanic sensors |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
11967127
|
YSI™
605913 |
Bottle of NaCl electrolyte, 1 sanding disc, six screw-on cap 1.25mL Polyethylene make DO membranes, yellow color |
71.70€
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Description
Specifications
Membrane Kit | |
For pro series galvanic sensors |
Product Title
YSI™ Pro Series DO Cap Membrane Kit