Learn More
YSI™ Pro Series DO Cap Membrane Kit
71.70€
Specifications
Type Membrane Kit
For Use With (Application) For pro series galvanic sensors
View More Specs
Products 1
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11967127
View Documents
YSI™
605913
Bottle of NaCl electrolyte, 1 sanding disc, six screw-on cap 1.25mL Polyethylene make DO membranes, yellow color
71.70€
Each
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
Specifications
Membrane Kit
For pro series galvanic sensors
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
YSI™ Pro Series DO Cap Membrane Kit