missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Pro Series DO Cap Membrane Kit

Product Code. 11967127
Click to view available options
Material:
Polyethylene Membrane
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11967127

Brand: YSI™ 605913

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Yellow
Type Membrane Kit
For Use With (Application) For pro series galvanic sensors
Includes Bottle of NaCl electrolyte, 1 sanding disc, six screw-on cap 1.25mL Polyethylene make DO membranes, yellow color
Material Polyethylene Membrane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.