Learn More
YSI™ DO Cap Membrane Kit
DO Cap Membrane Kit for use with probes and electrodes.
Brand: YSI™ 059880
Description
- DO Cap Membrane Kit
- For use with sensors and probes
Specifications
Instrument: 85, 550, and 556 MPS. Probes: 5239, 5905, 5010 and Pro Series Polarographic Sensors | |
12 in./sec. | |
PTFE |
DO Cap Membrane Kit | |
Black Cap Membrane, 1.0 mil | |
Oxygen/Temperature |