Learn More
Kern Sohn Bench Scale GAB
Compact bench scale that is practical for small spaces.
Brand:  Kern Sohn GAB 30K0.2N
Encompass_Preferred
 View more versions of this product
Product Code. 17301663
  • 402.90€ / Each
Explore more special offers
Description
Description
  • Compact size, practical for small spaces
  • High mobility: thanks to rechargeable battery operation (optional), compact, lightweight construction, it is suitable for the use in several locations (production, warehouse, dispatch department etc.)
  • Weighing with tolerance range (check-weighing): a visual and audible signal helps with portioning, dispensing or grading
  • Totalising of weights
  • Protective working cover included with delivery
Specifications
Specifications
30 kg
0.2 g
Compact
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Bench Scale GAB