Learn More
Kern Sohn Bench Scale GAB
Compact bench scale that is practical for small spaces.
POA - 402.90€
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
17381653
![]() |
Kern Sohn
GAB 12K0.1N |
12 kg |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17391653
![]() |
Kern Sohn
GAB 15K2DNM |
6 kg, 15 kg |
316.20€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17301663
![]() |
Kern Sohn
GAB 30K0.2N |
30 kg |
402.90€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17311663
![]() |
Kern Sohn
GAB 6K0.05N |
6 kg |
View alternative products
Save Up to » |
||||||
17321663
![]() |
Kern Sohn
GAB 6K1DNM |
3 kg, 6 kg |
316.20€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17240087
![]() |
Kern Sohn
GAB30K5DNM |
15 kg, 30 kg |
316.20€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Compact size, practical for small spaces
- High mobility: thanks to rechargeable battery operation (optional), compact, lightweight construction, it is suitable for the use in several locations (production, warehouse, dispatch department etc.)
- Weighing with tolerance range (check-weighing): a visual and audible signal helps with portioning, dispensing or grading
- Totalising of weights
- Protective working cover included with delivery
Specifications
Compact |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Bench Scale GAB