Learn More
Tuttnauer Wire (Mesh) Basket
Stainless steel mesh wire basket for bottles and general use.
Brand:  Tuttnauer BSK830-0003
 View more versions of this product
Product Code. 17869588
  • 140.00€ / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Description
  • Stainless Steel wire basket for the sterilization of solid items, or liquid in bottles.
  • The baskets are stackable and two baskets can be placed in the autoclave chamber.
Specifications
Specifications
5.7 gal.
25 cm
14.3 in.
Autoclave Basket
Non-sterile
26 L
10 in.
36.2 cm
362 x 250 mm
Product Title
Tuttnauer Wire (Mesh) Basket > Mesh Wire Basket; Capacity: 26 L (5.7 gal.); Dimensions (dia. x H): 362 x 250 mm; Material: Stainless Steel