Learn More
Socorex™ White Capillary Tubes
Make a connection to laboratory pipettes with Socorex™ White Capillary Tubes. Each tube is supplied in a practical box with a lid.
88.90€
Specifications
Sterility Sterile
Color White
Quantity 200/Pk.
Product Type Capillary Tube
Disposable Single-use
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10548442
View Documents
Socorex™
341.005
1 to 5 μL
88.90€
Pack of 200
In Stock
to see stock.
Edge
Add to basket
10330221
View Documents
Socorex™
341.025
5 to 25 μL
88.90€
Pack of 200
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Description
Description
  • White
  • Replacement glass capillaries
  • For Acura™ 846 positive displacement pipettes
  • Color coded to match corresponding pipette
Specifications
Specifications
Sterile
200/Pk.
Single-use
Glass
White
Capillary Tube
Acura 846 Positive Displacement Pipette
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Socorex™ White Capillary Tubes